Gene Transcripts

Gene Symbol
KRT28
Entrez Gene ID
162605
Gene Name
keratin 28
Gene Aliases
K25IRS4, KRT25D
Chromosome Location
Chr. 17: 40792196 - 40799959 on Build GRCh38
UniGene ID
Hs.59736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181535.3 NP_853513.2 8 1435
GenBank mRNA AJ564207.1 - 8 1408
AK129827.1 - 8 1435

View Full Product Details