Gene Transcripts

Gene Symbol
PARD3B
Entrez Gene ID
117583
Gene Name
par-3 family cell polarity regulator beta
Gene Aliases
ALS2CR19, PAR3B, PAR3L, PAR3beta
Chromosome Location
Chr. 2: 204545793 - 205620162 on Build GRCh38
UniGene ID
Hs.657382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302769.1 NP_001289698.1 12 1904
NM_057177.6 NP_476518.4 12 1904
NM_152526.5 NP_689739.4 11 1718
NM_205863.3 NP_995585.2 12 1904
XM_011510552.2 XP_011508854.1 15 2247
XM_011510553.2 XP_011508855.1 14 6970
XM_017003283.1 XP_016858772.1 11 2194
XM_017003284.1 XP_016858773.1 12 5240
XM_017003285.1 XP_016858774.1 13 6791
XM_017003286.1 XP_016858775.1 11 5136
XM_017003287.1 XP_016858776.1 14 6973
XM_017003288.1 XP_016858777.1 14 6975
XM_017003289.1 XP_016858778.1 9 1351
XM_017003290.1 XP_016858779.1 13 1851
XM_017003291.1 XP_016858780.1 12 1723
XM_017003292.1 XP_016858781.1 12 1785
XM_017003293.1 XP_016858782.1 9 1289
XM_017003294.1 XP_016858783.1 14 6967
GenBank mRNA AB073472.1 - 14 1697
AB092439.1 - 14 1697
AF428250.1 - 12 1697
AF428251.1 - 11 1511
AF466152.1 - 12 1697
AK056157.1 - 14 1785
AK057965.1 - 7 1062

View Full Product Details