Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166412.1 | NP_001159884.1 | 13 | 1564 | |
NM_022138.2 | NP_071421.1 | 13 | 1597 | ||
GenBank mRNA | AB014730.1 | - | 13 | 1364 | |
AB014737.1 | - | 13 | 1397 | ||
AF173892.1 | - | 12 | 1219 | ||
AF176778.1 | - | 3 | 329 | ||
AJ249902.1 | - | 8 | 843 | ||
AJ420521.1 | - | 13 | 1544 | ||
AK056700.1 | - | 13 | 1491 | ||
AK297839.1 | - | 5 | 489 | ||
AK315920.1 | - | 5 | 853 | ||
AL832303.1 | - | 13 | 1499 | ||
BC028420.1 | - | 13 | 1567 | 4826774 | |
BC047583.1 | - | 13 | 1536 | 4826950 |