Gene Transcripts

Gene Symbol
LRRC2
Entrez Gene ID
79442
Gene Name
leucine rich repeat containing 2
Gene Aliases
Chromosome Location
Chr. 3: 46515385 - 46580099 on Build GRCh38
UniGene ID
Hs.657345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024512.4 NP_078788.2 3 694
XM_011534110.2 XP_011532412.1 3 483
XM_017007177.1 XP_016862666.1 3 694
GenBank mRNA AJ308569.2 - 3 468
AK057808.1 - 3 536
AK223440.1 - 3 536
AK314658.1 - 3 457
AK315636.1 - 3 477
BC029118.1 - 3 466 5183639

View Full Product Details