Gene Transcripts
- Gene Symbol
- CEP76
- Entrez Gene ID
- 79959
- Gene Name
- centrosomal protein 76
- Gene Aliases
- C18orf9, HsT1705
- Chromosome Location
- Chr. 18: 12661733 - 12702777 on Build GRCh38
- UniGene ID
- Hs.236940
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271989.1 |
NP_001258918.1
|
8, 9 |
1301 |
|
NM_024899.3 |
NP_079175.2
|
9, 10 |
1526 |
|
NR_073537.1 |
|
9, 10 |
1521 |
|
XM_005258149.4 |
XP_005258206.1
|
9, 10 |
1505 |
|
XM_017025981.1 |
XP_016881470.1
|
9, 10 |
1505 |
|
XM_017025982.1 |
XP_016881471.1
|
9, 10 |
1586 |
|
XM_017025983.1 |
XP_016881472.1
|
8, 9 |
1376 |
|
XM_017025984.1 |
XP_016881473.1
|
9, 10 |
1488 |
|
XM_017025985.1 |
XP_016881474.1
|
9, 10 |
1454 |
|
XM_017025986.1 |
XP_016881475.1
|
8, 9 |
1263 |
|
XM_017025987.1 |
XP_016881476.1
|
8, 9 |
1230 |
|
GenBank mRNA |
AK022604.1 |
- |
9, 10 |
1388 |
|
AK303117.1 |
- |
8, 9 |
1248 |
|
BC026307.1 |
- |
9, 10 |
1453 |
4792211 |
BC033026.1 |
- |
9, 10 |
1388 |
|
View Full Product Details