Gene Transcripts

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
Chromosome Location
Chr. 1: 224385143 - 224434299 on Build GRCh38
UniGene ID
Hs.497873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115113.2 NP_001108585.2 14 3558
NM_025160.6 NP_079436.4 14 3606
XM_005273273.4 XP_005273330.1 13 3321
XM_006711817.3 XP_006711880.1 14 3504
XM_011544281.2 XP_011542583.1 14 3347
XM_011544282.2 XP_011542584.1 11 2669
XM_017002396.1 XP_016857885.1 14 3195
GenBank mRNA AB209887.1 - 14 3606
AK027236.1 - 1 812
AY221751.1 - 14 3396
AY304473.1 - 14 3163
BC031471.2 - 9 2549
BC063817.1 - 10 2666

View Full Product Details