Gene Transcripts

Gene Symbol
TRIM8
Entrez Gene ID
81603
Gene Name
tripartite motif containing 8
Gene Aliases
GERP, RNF27
Chromosome Location
Chr. 10: 102644484 - 102658319 on Build GRCh38
UniGene ID
Hs.336810
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030912.2 NP_112174.2 3 1009
GenBank mRNA AF220034.1 - 3 886
AF281046.1 - 3 895
BC021925.1 - 3 1008 4857393

View Full Product Details