Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
89822
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
K2p17.1, TALK-2, TALK2, TASK-4, TASK4
Chromosome Location
Chr. 6: 39299001 - 39314461 on Build GRCh38
UniGene ID
Hs.162282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135111.1 NP_001128583.1 6 1652
NM_031460.3 NP_113648.2 5 1500
GenBank mRNA AY358853.1 - 5 1451
BC025726.1 - 5 1447 5201326

View Full Product Details