Gene Transcripts

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
Chromosome Location
Chr. 6: 145798135 - 145864409 on Build GRCh38
UniGene ID
Hs.421095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032145.4 NP_115521.3 2 1210
XM_005267159.3 XP_005267216.1 2 1049
XM_011536177.2 XP_011534479.1 2 6069
XM_017011353.1 XP_016866842.1 2 1049
GenBank mRNA AF248640.1 - 1 91
AK124138.1 - 1 410
BC024326.1 - 2 1178 3915249

View Full Product Details