Gene Transcripts

Gene Symbol
LSM10
Entrez Gene ID
84967
Gene Name
LSM10, U7 small nuclear RNA associated
Gene Aliases
MST074, MSTP074
Chromosome Location
Chr. 1: 36393420 - 36397959 on Build GRCh38
UniGene ID
Hs.603131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032881.2 NP_116270.1 2 710
GenBank mRNA AF166092.1 - 2 636
AF394685.1 - 2 608
BC007623.1 - 2 643 3357043

View Full Product Details