Gene Transcripts

Gene Symbol
TNKS1BP1
Entrez Gene ID
85456
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
TAB182
Chromosome Location
Chr. 11: 57299629 - 57324952 on Build GRCh38
UniGene ID
Hs.530730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033396.2 NP_203754.2 6 3848
XM_006718725.3 XP_006718788.1 6 3864
XM_011545325.2 XP_011543627.1 6 3861
GenBank mRNA AB051528.2 - 6 3828
AF441771.1 - 5 3990
AK074113.1 - 2 2205
AK124903.1 - 6 3875
AK127871.1 - 4 3271
BC150333.1 - 6 3839 8860508

View Full Product Details