Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080593.2 | NP_542160.1 | 2 | 599 | |
GenBank mRNA | AJ223352.1 | - | 2 | 555 | |
BC000893.1 | - | 2 | 581 | 3463930 | |
BC051872.1 | - | 2 | 579 | 6526835 | |
BC064959.1 | - | 2 | 579 | 6093977 | |
BC108737.1 | - | 2 | 579 | 4707078 |