Gene Transcripts

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
Chromosome Location
Chr. 9: 92456205 - 92482562 on Build GRCh38
UniGene ID
Hs.435655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193335.1 NP_001180264.1 6 1373
NM_017680.4 NP_060150.4 8 1606
GenBank mRNA AK000136.1 - 6 976
AK027359.1 - 6 1176
AK172737.1 - 6 1076
AK304501.1 - 6 1180
AY029191.1 - 8 1539
AY358329.1 - 8 1539
BC063114.1 - 8 1571 30404602

View Full Product Details