Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318485.1 | NP_001305414.1 | 4 | 745 | |
NM_017971.3 | NP_060441.2 | 4 | 400 | ||
GenBank mRNA | AB049644.1 | - | 4 | 331 | |
AK000886.1 | - | 3 | 1356 | ||
AK315794.1 | - | 4 | 334 | ||
BC009515.1 | - | 4 | 368 | 3542715 | |
BC014316.1 | - | 4 | 337 | 4045076 | |
BC059945.1 | - | 4 | 318 | 5397911 | |
BG700526.1 | - | 4 | 400 | ||
BM818788.1 | - | 3 | 542 |