Gene Transcripts

Gene Symbol
TMEM30A
Entrez Gene ID
55754
Gene Name
transmembrane protein 30A
Gene Aliases
C6orf67, CDC50A
Chromosome Location
Chr. 6: 75252922 - 75284916 on Build GRCh38
UniGene ID
Hs.108530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143958.1 NP_001137430.1 2 568
NM_018247.3 NP_060717.1 3 676
GenBank mRNA AK001718.1 - 3 545
AK292823.1 - 2 433
AK293225.1 - 4 501
AK295567.1 - 4 497
AL832490.1 - 3 599
AL832815.1 - 3 538
BC009006.1 - 2 419 4154200

View Full Product Details