Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018283.3 | NP_060753.1 | 3 | 669 | |
NR_136687.1 | 3 | 669 | |||
GenBank mRNA | AK001818.1 | - | 3 | 669 | |
BC064607.1 | - | 3 | 488 | ||
BC107875.1 | - | 3 | 542 | 6451308 | |
BC133015.1 | - | 3 | 610 | 40147037 | |
BC133017.1 | - | 3 | 610 | 40147039 | |
DA000395.1 | - | 3 | 454 | ||
DA218525.1 | - | 1 | 54 |