Gene Transcripts

Gene Symbol
AP5S1
Entrez Gene ID
55317
Gene Name
adaptor related protein complex 5 sigma 1 subunit
Gene Aliases
C20orf29
Chromosome Location
Chr. 20: 3820524 - 3825307 on Build GRCh38
UniGene ID
Hs.104806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204446.1 NP_001191375.1 3 535
NM_001204447.1 NP_001191376.1 3 383
NM_018347.2 NP_060817.1 3 439
GenBank mRNA AK002030.1 - 3 407
AK093332.1 - 3 351
BC043344.1 - 3 376 5505030
DA129391.1 - 3 503
DA253969.1 - 3 439
DB190907.1 - 3 383

View Full Product Details