Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098536.1 | NP_001092006.1 | 4 | 421 | |
NM_003481.2 | NP_003472.2 | 4 | 415 | ||
GenBank mRNA | AK308021.1 | - | 4 | 405 | |
BC004889.2 | - | 4 | 394 | 3936781 | |
BC005139.2 | - | 4 | 392 | 3506801 | |
CN276110.1 | - | 4 | 415 | ||
U35116.1 | - | 4 | 405 | ||
U47927.1 | - | 4 | 414 | ||
X91349.1 | - | 4 | 391 |