Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr. 14: 88385638 - 88470350 on Build GRCh38
UniGene ID
Hs.525518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040428.3 NP_001035518.1 7, 8 1115
NM_018418.4 NP_060888.2 8, 9 1211
XM_005267851.1 XP_005267908.1 8, 9 1185
XM_005267852.1 XP_005267909.1 7, 8 1114
XM_005267854.1 XP_005267911.1 9, 10 1247
XM_005267855.1 XP_005267912.1 8, 9 1180
XM_006720204.1 XP_006720267.1 8, 9 1185
XM_006720205.1 XP_006720268.1 8, 9 1185
XM_011536951.1 XP_011535253.1 8, 9 1379
XM_011536952.1 XP_011535254.1 6, 7 997
XM_011536953.1 XP_011535255.1 7, 8 1318
XM_017021452.1 XP_016876941.1 8, 9 1376
XM_017021453.1 XP_016876942.1 8, 9 1345
XM_017021454.1 XP_016876943.1 9, 10 1244
XM_017021455.1 XP_016876944.1 8, 9 1177
XM_017021456.1 XP_016876945.1 9, 10 1190
XM_017021457.1 XP_016876946.1 7, 8 1315
GenBank mRNA AF144487.3 - 8, 9 1191
AF144488.2 - 6, 7 919
AK055864.1 - 7, 8 2783
AK225746.1 - 9, 10 1185
AK290631.1 - 7, 8 1086
AL136604.1 - 7, 8 1095
BC008656.1 - 3, 4 643
BC090875.1 - 8, 9 1498 30383306
CR533563.1 - 7, 8 940
CR936777.1 - 8, 9 1157
EU668353.1 - 8, 9 1187

View Full Product Details