Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 3 | 702 | |
XM_011535926.1 | XP_011534228.1 | 3 | 702 | ||
XM_011535927.2 | XP_011534229.1 | 2 | 159 | ||
XM_011535928.2 | XP_011534230.1 | 2 | 348 | ||
XM_017011003.1 | XP_016866492.1 | 3 | 370 | ||
GenBank mRNA | AJ278110.1 | - | 3 | 565 | |
AK303324.1 | - | 3 | 411 | ||
AK313686.1 | - | 3 | 402 | ||
AL136751.1 | - | 3 | 390 | ||
BC066938.1 | - | 3 | 383 | 4829072 | |
BX646425.1 | - | 3 | 449 |