Gene Transcripts

Gene Symbol
MIS18A
Entrez Gene ID
54069
Gene Name
MIS18 kinetochore protein A
Gene Aliases
B28, C21orf45, C21orf46, FASP1, MIS18alpha, hMis18alpha
Chromosome Location
Chr. 21: 32244759 - 32283954 on Build GRCh38
UniGene ID
Hs.190518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018944.2 NP_061817.1 5 1452
GenBank mRNA AK075281.1 - 5 1426

View Full Product Details