Gene Transcripts

Gene Symbol
TOLLIP
Entrez Gene ID
54472
Gene Name
toll interacting protein
Gene Aliases
IL-1RAcPIP
Chromosome Location
Chr. 11: 1274368 - 1309666 on Build GRCh38
UniGene ID
Hs.368527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318512.1 NP_001305441.1 5 2924
NM_001318514.1 NP_001305443.1 6 3084
NM_001318515.1 NP_001305444.1 3 2647
NM_001318516.1 NP_001305445.1 5 2891
NM_019009.3 NP_061882.2 6 3074
XM_017017931.1 XP_016873420.1 4 2981
GenBank mRNA AB209541.1 - 7 2779
AK096693.1 - 5 2849
AK127123.1 - 6 3038
AL136835.1 - 6 3021
BX648136.1 - 1 3351

View Full Product Details