Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286238.1 | NP_001273167.1 | 5 | 524 | |
NM_001286239.1 | NP_001273168.1 | 4 | 482 | ||
NM_019073.3 | NP_061946.1 | 5 | 524 | ||
XM_006710699.2 | XP_006710762.1 | 5 | 518 | ||
XM_006710701.3 | XP_006710764.1 | 5 | 518 | ||
XM_011541606.1 | XP_011539908.1 | 5 | 518 | ||
XM_011541608.1 | XP_011539910.1 | 4 | 461 | ||
XM_011541609.2 | XP_011539911.1 | 3 | 169 | ||
XM_017001509.1 | XP_016856998.1 | 5 | 518 | ||
XM_017001511.1 | XP_016857000.1 | 5 | 518 | ||
XM_017001512.1 | XP_016857001.1 | 6 | 367 | ||
XM_017001513.1 | XP_016857002.1 | 3 | 131 | ||
XM_017001514.1 | XP_016857003.1 | 5 | 345 | ||
XM_017001515.1 | XP_016857004.1 | 5 | 346 | ||
GenBank mRNA | AK000869.1 | - | 5 | 489 | |
AK301770.1 | - | 4 | 425 | ||
BC020660.1 | - | 5 | 326 | 4657425 |