Gene Transcripts

Gene Symbol
UBQLN4
Entrez Gene ID
56893
Gene Name
ubiquilin 4
Gene Aliases
A1U, A1Up, C1orf6, CIP75, UBIN
Chromosome Location
Chr. 1: 156035294 - 156053825 on Build GRCh38
UniGene ID
Hs.283739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304342.1 NP_001291271.1 4 705
NM_020131.4 NP_064516.2 4 765
XM_005245348.3 XP_005245405.1 4 670
GenBank mRNA AB209566.1 - 4 638
AF113544.1 - 1 171
AF188240.1 - 4 919
AK302895.1 - 4 594
AK314413.1 - 4 653
BC018403.2 - 4 653 4214386
BC063841.1 - 4 665 6183942

View Full Product Details