Gene Transcripts

Gene Symbol
C5orf15
Entrez Gene ID
56951
Gene Name
chromosome 5 open reading frame 15
Gene Aliases
HTGN29, KCT2
Chromosome Location
Chr. 5: 133955507 - 133968715 on Build GRCh38
UniGene ID
Hs.744034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020199.2 NP_064584.1 3 1686
GenBank mRNA AF035313.1 - 1 778
AF226055.1 - 3 1762
AK074963.1 - 3 1598
AK298515.1 - 4 1495
BC020875.1 - 3 1577 4610607

View Full Product Details