Gene Transcripts

Gene Symbol
PLXDC1
Entrez Gene ID
57125
Gene Name
plexin domain containing 1
Gene Aliases
TEM3, TEM7
Chromosome Location
Chr. 17: 39063303 - 39151649 on Build GRCh38
UniGene ID
Hs.125036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020405.4 NP_065138.2 14 2194
GenBank mRNA AF279144.2 - 14 2064
AF378753.1 - 14 2064
AK093589.1 - 13 2493
AK127539.1 - 13 2375
AK303699.1 - 13 1881
AL833517.1 - 6 1070
AY704670.1 - 14 2428
AY704671.1 - 15 2628
AY704672.1 - 14 2374
BC036059.1 - 14 2160 5298499

View Full Product Details