Gene Transcripts

Gene Symbol
PLEKHB1
Entrez Gene ID
58473
Gene Name
pleckstrin homology domain containing B1
Gene Aliases
KPL1, PHR1, PHRET1
Chromosome Location
Chr. 11: 73646178 - 73662819 on Build GRCh38
UniGene ID
Hs.445489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130033.1 NP_001123505.1 7 2161
NM_001130034.1 NP_001123506.1 8 2007
NM_001130035.1 NP_001123507.1 7 1902
NM_001130036.1 NP_001123508.1 6 2034
NM_021200.2 NP_067023.1 8 2266
XM_011545192.2 XP_011543494.1 8 2840
XM_011545193.2 XP_011543495.1 7 2735
GenBank mRNA AB209716.1 - 7 1814
AF081583.1 - 7 1784
AF093249.1 - 7 1786
AF100612.1 - 8 1891
AF101054.1 - 7 2157
AK054725.1 - 3 1919
BC008075.2 - 7 1840 3050369
CK819503.1 - 1 441
U89715.1 - 8 2262

View Full Product Details