Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136039.2 | NP_001129511.1 | 4 | 721 | |
NM_001142355.1 | NP_001135827.1 | 4 | 670 | ||
NM_001142356.1 | NP_001135828.1 | 3 | 656 | ||
NM_021824.3 | NP_068596.2 | 4 | 873 | ||
XM_005246742.3 | XP_005246799.1 | 4 | 936 | ||
XM_011511582.1 | XP_011509884.1 | 4 | 857 | ||
XM_011511583.1 | XP_011509885.1 | 4 | 1601 | ||
XM_011511584.1 | XP_011509886.1 | 4 | 1419 | ||
XM_011511585.1 | XP_011509887.1 | 4 | 1368 | ||
XM_017004630.1 | XP_016860119.1 | 4 | 1118 | ||
GenBank mRNA | AB038949.1 | - | 4 | 834 | |
AF060513.1 | - | 4 | 630 | ||
AF182416.1 | - | 4 | 795 | ||
AF283538.1 | - | 4 | 676 | ||
AK023378.1 | - | 4 | 819 | ||
AK225905.1 | - | 4 | 2196 | ||
AK314709.1 | - | 4 | 675 | ||
AL834430.1 | - | 3 | 496 | ||
AY251943.1 | - | 3 | 659 | ||
AY251944.1 | - | 3 | 659 | ||
BC007654.2 | - | 3 | 657 | 3346099 |