Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003654.2 | 3 | 943 | ||
NR_004859.1 | 4 | 1501 | |||
GenBank mRNA | AA280726.1 | - | 3 | 205 | |
AF229246.1 | - | 4 | 2761 | ||
AI218957.1 | - | 3 | 205 | ||
AK022844.1 | - | 3 | 943 | ||
AK290489.1 | - | 3 | 953 | ||
AL109690.1 | - | 3 | 1501 | ||
AW341587.1 | - | 4 | 285 | ||
BC011547.1 | - | 3 | 925 | ||
BC012929.1 | - | 3 | 925 | ||
BC063836.1 | - | 3 | 899 |