Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022728.3 | NP_073565.2 | 2 | 2085 | |
GenBank mRNA | AF136169.1 | - | 2 | 1852 | |
AF248954.1 | - | 2 | 2085 | ||
AK055238.1 | - | 2 | 2055 | ||
BC035048.2 | - | 2 | 1887 | 5261681 |