Gene Transcripts
- Gene Symbol
- SCOC
- Entrez Gene ID
- 60592
- Gene Name
- short coiled-coil protein
- Gene Aliases
- HRIHFB2072, SCOCO, UNC-69
- Chromosome Location
- Chr. 4: 140257286 - 140382556 on Build GRCh38
- UniGene ID
- Hs.480815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001153446.1 |
NP_001146918.1
|
2, 3 |
125 |
|
NM_001153484.1 |
NP_001146956.1
|
1, 2 |
218 |
|
NM_001153552.1 |
NP_001147024.1
|
1, 2 |
218 |
|
NM_001153585.1 |
NP_001147057.1
|
1, 2 |
258 |
|
NM_001153635.1 |
NP_001147107.1
|
2, 3 |
125 |
|
NM_001153663.1 |
NP_001147135.1
|
1, 2 |
218 |
|
NM_001153690.1 |
NP_001147162.1
|
1, 2 |
258 |
|
NM_032547.2 |
NP_115936.2
|
2, 3 |
223 |
|
GenBank mRNA |
AB015335.1 |
- |
1, 2 |
89 |
|
AF330205.1 |
- |
1, 2 |
109 |
|
AF448857.1 |
- |
1, 2 |
109 |
|
AK027797.1 |
- |
2, 3 |
223 |
|
BC016511.1 |
- |
1, 2 |
60 |
|
BC062684.1 |
- |
2, 3 |
125 |
4691440 |
BU070009.1 |
- |
1, 2 |
197 |
|
CN367654.1 |
- |
1, 2 |
218 |
|
DA625782.1 |
- |
1, 2 |
258 |
|
DA695503.1 |
- |
1, 2 |
192 |
|
DB204055.1 |
- |
2, 3 |
125 |
|
DB465370.1 |
- |
1, 2 |
167 |
|
View Full Product Details