Gene Transcripts

Gene Symbol
DMRTC1
Entrez Gene ID
63947
Gene Name
DMRT like family C1
Gene Aliases
Chromosome Location
Chr. X: 72872025 - 72943653 on Build GRCh38
UniGene ID
Hs.558685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033053.2 NP_149042.2 6 709
XM_005262287.3 XP_005262344.1 7 788
XM_005262288.3 XP_005262345.1 7 813
XM_017029725.1 XP_016885214.1 7 819
GenBank mRNA AV753283.1 - 254
AW205069.1 - 2 269
BC029799.1 - 6 1133
BC047596.1 - 5 657 4830694

Gene Symbol
DMRTC1B
Entrez Gene ID
728656
Gene Name
DMRT like family C1B
Gene Aliases
Chromosome Location
Chr. X: 72777170 - 72848802 on Build GRCh38
UniGene ID
Hs.712728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080851.1 NP_001074320.1 6 709
XM_006724684.2 XP_006724747.1 7 788
XM_006724685.3 XP_006724748.1 7 813
XM_017029776.1 XP_016885265.1 7 819

View Full Product Details