Gene Transcripts

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
Chromosome Location
Chr. 2: 162318768 - 162371605 on Build GRCh38
UniGene ID
Hs.377894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012198.3 NP_036330.1 2 278
XM_005246446.2 XP_005246503.1 2 219
XM_006712398.3 XP_006712461.1 2 451
XM_006712400.3 XP_006712463.1 3 529
XM_006712401.3 XP_006712464.1 2 530
XM_011510926.2 XP_011509228.1 2 230
XM_011510927.2 XP_011509229.1 2 225
XM_017003764.1 XP_016859253.1 3 542
XM_017003765.1 XP_016859254.1 3 628
XM_017003766.1 XP_016859255.1 3 425
XM_017003767.1 XP_016859256.1 2 301
XM_017003768.1 XP_016859257.1 2 296
XM_017003769.1 XP_016859258.1 2 451
GenBank mRNA AB209768.1 - 2 167
AK295413.1 - 2 211
AK312349.1 - 2 227
BC005214.1 - 2 201 3996889
M81637.1 - 2 221

View Full Product Details