Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013313.4 | NP_037445.1 | 5 | 2240 | |
NR_130910.1 | 5 | 2624 | |||
GenBank mRNA | AB098734.1 | - | 5 | 2237 | |
AK096530.1 | - | 1 | 102 | ||
BC034486.1 | - | 5 | 2099 | 4826997 | |
BI836931.1 | - | 1 | 102 | ||
CR933630.1 | - | 4 | 6344 |