Gene Transcripts

Gene Symbol
STAU2
Entrez Gene ID
27067
Gene Name
staufen double-stranded RNA binding protein 2
Gene Aliases
39K2, 39K3
Chromosome Location
Chr. 8: 73420369 - 73747708 on Build GRCh38
UniGene ID
Hs.561815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164380.1 NP_001157852.1 5 592
NM_001164381.1 NP_001157853.1 4 461
NM_001164384.1 NP_001157856.1 3 525
NM_001164385.1 NP_001157857.1 2 282
NM_014393.2 NP_055208.2 4 461
GenBank mRNA AB210014.1 - 4 357
AF459097.1 - 3 311
AF459098.1 - 2 180
AK002152.1 - 5 505
AK293729.1 - 2 222
AK294466.1 - 2 234
AK314617.1 - 4 374
AL079285.1 - 4 384
BC008369.1 - 4 374 3682739
BC008370.1 - 4 374
BC110447.1 - 2 198 40009098
DB501161.1 - 2 282
Y19062.1 - 4 356

View Full Product Details