Gene Transcripts

Gene Symbol
DKK2
Entrez Gene ID
27123
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Gene Aliases
DKK-2
Chromosome Location
Chr. 4: 106921802 - 107036296 on Build GRCh38
UniGene ID
Hs.211869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014421.2 NP_055236.1 4 3081
GenBank mRNA AF177395.1 - 4 3093
AY358414.1 - 4 2586

View Full Product Details