Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014521.2 | NP_055336.1 | 4 | 2819 | |
XM_011510891.2 | XP_011509193.1 | 4 | 2839 | ||
XM_011510892.1 | XP_011509194.1 | 4 | 2828 | ||
XM_011510893.1 | XP_011509195.1 | 4 | 2753 | ||
XM_011510894.2 | XP_011509196.1 | 4 | 5252 | ||
GenBank mRNA | AF147747.1 | - | 4 | 2761 | |
AK291209.1 | - | 4 | 2786 | ||
AK304398.1 | - | 2 | 921 | ||
BC057396.1 | - | 4 | 2830 | 6138465 | |
BX647198.1 | - | 4 | 2746 | ||
DQ232895.1 | - | 4 | 2793 |