Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001326578.1 | NP_001313507.1 | 8, 9 | 933 | |
NM_001326579.1 | NP_001313508.1 | 8, 9 | 720 | ||
NM_001326580.1 | NP_001313509.1 | 8, 9 | 856 | ||
NM_003569.2 | NP_003560.2 | 8, 9 | 856 | ||
XM_011536178.1 | XP_011534480.1 | 8, 9 | 772 | ||
GenBank mRNA | AK294911.1 | - | 8, 9 | 605 | |
BC011975.1 | - | 8, 9 | 720 | 3860059 | |
U77942.1 | - | 8, 9 | 685 |