Gene Transcripts

Gene Symbol
UPF2
Entrez Gene ID
26019
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
HUPF2, RENT2, smg-3
Chromosome Location
Chr. 10: 11920022 - 12043170 on Build GRCh38
UniGene ID
Hs.370689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015542.3 NP_056357.1 20 3755
NM_080599.2 NP_542166.1 20 3925
XM_017016034.1 XP_016871523.1 19 3411
GenBank mRNA AB037829.1 - 19 4124
AF301013.1 - 20 3748
AF318574.1 - 20 3725
AK000764.1 - 5 586
AK074375.1 - 5 519
AK295209.1 - 7 824
AL080198.1 - 14 1925
AY013249.1 - 19 3674
BC114964.1 - 20 3719 40082739
BC115737.1 - 20 3719 40082728

View Full Product Details