Gene Transcripts

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
Chromosome Location
Chr. 19: 9959561 - 10010532 on Build GRCh38
UniGene ID
Hs.235368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015719.3 NP_056534.2 67 5202
XM_011528042.2 XP_011526344.1 67 5260
XM_017026849.1 XP_016882338.1 40 2903
GenBank mRNA AB209175.1 - 65 4855
AF177941.1 - 67 5202

View Full Product Details