Gene Transcripts

Gene Symbol
SLC35B3
Entrez Gene ID
51000
Gene Name
solute carrier family 35 member B3
Gene Aliases
C6orf196, CGI-19, PAPST2
Chromosome Location
Chr. 6: 8413066 - 8435606 on Build GRCh38
UniGene ID
Hs.285847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142541.2 NP_001136013.1 10 1190
NM_015948.4 NP_057032.2 10 1552
NR_109913.1 11 1543
NR_109914.1 8 1486
XM_005249156.3 XP_005249213.1 9 1332
XM_005249157.2 XP_005249214.1 10 1325
XM_005249158.1 XP_005249215.1 9 1220
XM_006715102.3 XP_006715165.1 10 1362
XM_006715103.3 XP_006715166.1 8 1239
XM_011514648.1 XP_011512950.1 11 1332
XM_017010910.1 XP_016866399.1 10 1648
XM_017010911.1 XP_016866400.1 9 1566
XM_017010912.1 XP_016866401.1 9 1187
XM_017010913.1 XP_016866402.1 9 1555
XM_017010914.1 XP_016866403.1 9 1416
XM_017010915.1 XP_016866404.1 8 1127
GenBank mRNA AB231931.1 - 8 1027
AF132953.1 - 9 1048
AK313522.1 - 10 1168
BC006973.1 - 10 1378 3827889
BX538271.1 - 11 1539
BX641086.1 - 9 6691
BX647208.1 - 8 1420
BX648022.1 - 9 5489

View Full Product Details