Gene Transcripts
- Gene Symbol
- LUC7L2
- Entrez Gene ID
- 51631
- Gene Name
- LUC7 like 2, pre-mRNA splicing factor
- Gene Aliases
- CGI-59, CGI-74, LUC7B2
- Chromosome Location
- Chr. 7: 139340359 - 139423457 on Build GRCh38
- UniGene ID
- Hs.731488
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001244585.1 |
NP_001231514.1
|
6, 7 |
693 |
|
NM_001270643.1 |
NP_001257572.1
|
6, 7 |
1005 |
|
NM_016019.4 |
NP_057103.2
|
5, 6 |
934 |
|
GenBank mRNA |
AF151817.1 |
- |
5, 6 |
493 |
|
AF151832.1 |
- |
5, 6 |
518 |
|
AK001476.1 |
- |
5, 6 |
826 |
|
AK022895.1 |
- |
5, 6 |
809 |
|
AK092044.1 |
- |
4, 5 |
1315 |
|
AK093864.1 |
- |
6, 7 |
792 |
|
AK095839.1 |
- |
4, 5 |
1846 |
|
AK298166.1 |
- |
6, 7 |
693 |
|
BC017163.2 |
- |
5, 6 |
851 |
3920377 |
BC042625.1 |
- |
6, 7 |
939 |
5178074 |
BC050708.2 |
- |
5, 6 |
864 |
6500791 |
BC056886.1 |
- |
5, 6 |
809 |
6147116 |
- Gene Symbol
- C7orf55-LUC7L2
- Entrez Gene ID
- 100996928
- Gene Name
- C7orf55-LUC7L2 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 7: 139341132 - 139423457 on Build GRCh38
- UniGene ID
- Hs.731488
- Species
- Homo sapiens
View Full Product Details