Gene Transcripts
- Gene Symbol
- SUCO
- Entrez Gene ID
- 51430
- Gene Name
- SUN domain containing ossification factor
- Gene Aliases
- C1orf9, CH1, OPT, SLP1
- Chromosome Location
- Chr. 1: 172532349 - 172611833 on Build GRCh38
- UniGene ID
- Hs.204559
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282750.1 |
NP_001269679.1
|
12, 13 |
1546 |
|
NM_001282751.1 |
NP_001269680.1
|
13, 14 |
1657 |
|
NM_014283.4 |
NP_055098.1
|
13, 14 |
1657 |
|
NM_016227.3 |
NP_057311.3
|
12, 13 |
1916 |
|
XM_006711374.2 |
XP_006711437.1
|
12, 13 |
1717 |
|
XM_006711375.2 |
XP_006711438.1
|
12, 13 |
1627 |
|
XM_006711376.1 |
XP_006711439.1
|
11, 12 |
1606 |
|
GenBank mRNA |
AF097535.1 |
- |
12, 13 |
1916 |
|
AJ250075.1 |
- |
13, 14 |
1655 |
|
AK302509.1 |
- |
12, 13 |
1481 |
|
AK302949.1 |
- |
13, 14 |
1657 |
|
AL035291.1 |
- |
13, 14 |
1558 |
|
BC137125.1 |
- |
13, 14 |
1443 |
9021122 |
BC144487.1 |
- |
13, 14 |
1443 |
|
View Full Product Details