Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017425.3 | NP_059121.1 | 3 | 342 | |
XM_011542870.2 | XP_011541172.1 | 3 | 532 | ||
GenBank mRNA | AF334735.1 | - | 3 | 341 | |
AK311806.1 | - | 3 | 285 | ||
BC019224.1 | - | 3 | 274 | ||
BC032457.2 | - | 2 | 234 | 5171849 | |
DA511916.1 | - | 3 | 391 | ||
KU257683.1 | - | 2 | 206 |