Gene Transcripts

Gene Symbol
SH3KBP1
Entrez Gene ID
30011
Gene Name
SH3 domain containing kinase binding protein 1
Gene Aliases
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
Chromosome Location
Chr. X: 19533965 - 19887626 on Build GRCh38
UniGene ID
Hs.726365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024666.2 NP_001019837.1 3 366
NM_031892.2 NP_114098.1 4 678
XM_005274494.2 XP_005274551.1 4 544
XM_011545498.2 XP_011543800.1 4 544
XM_011545499.2 XP_011543801.1 3 352
XM_011545500.2 XP_011543802.1 4 544
XM_011545503.2 XP_011543805.1 4 544
XM_017029459.1 XP_016884948.1 3 284
XM_017029460.1 XP_016884949.1 3 327
XM_017029461.1 XP_016884950.1 3 327
XM_017029462.1 XP_016884951.1 4 544
XM_017029463.1 XP_016884952.1 3 386
XM_017029464.1 XP_016884953.1 4 619
XM_017029465.1 XP_016884954.1 4 645
XM_017029466.1 XP_016884955.1 4 544
XM_017029467.1 XP_016884956.1 4 618
XM_017029468.1 XP_016884957.1 4 645
GenBank mRNA AF230904.1 - 4 653
AF542051.1 - 3 329
AK293234.1 - 3 306
AK313308.1 - 4 606
AY423734.1 - 5 576
AY542305.1 - 4 413
BC015806.1 - 4 580 3906722
BC050663.2 - 4 588
DC409010.1 - 3 366
DR000949.1 - 4 705

View Full Product Details