Gene Transcripts
- Gene Symbol
- SCML2
- Entrez Gene ID
- 10389
- Gene Name
- sex comb on midleg-like 2 (Drosophila)
- Gene Aliases
- Chromosome Location
- Chr. X: 18239313 - 18355089 on Build GRCh38
- UniGene ID
- Hs.495774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006089.2 |
NP_006080.1
|
5, 6 |
553 |
|
NR_033717.1 |
|
5, 6 |
553 |
|
XM_006724458.1 |
XP_006724521.1
|
5, 6 |
517 |
|
XM_017029219.1 |
XP_016884708.1
|
5, 6 |
449 |
|
XM_017029220.1 |
XP_016884709.1
|
5, 6 |
445 |
|
XM_017029221.1 |
XP_016884710.1
|
5, 6 |
587 |
|
GenBank mRNA |
AK303064.1 |
- |
4, 5 |
419 |
|
AK315116.1 |
- |
5, 6 |
517 |
|
BC051913.1 |
- |
5, 6 |
495 |
|
BC064617.1 |
- |
5, 6 |
510 |
5575894 |
Y18004.1 |
- |
5, 6 |
487 |
|
View Full Product Details