Gene Transcripts

Gene Symbol
EBNA1BP2
Entrez Gene ID
10969
Gene Name
EBNA1 binding protein 2
Gene Aliases
EBP2, NOBP, P40
Chromosome Location
Chr. 1: 43164174 - 43172570 on Build GRCh38
UniGene ID
Hs.346868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159936.1 NP_001153408.1 6 836
NM_006824.2 NP_006815.2 5 719
GenBank mRNA AB451256.1 - 5 522
AB451381.1 - 5 522
AK223455.1 - 5 702
AK295006.1 - 4 1305
AK307440.1 - 5 665
BC009175.2 - 5 660 2966681
BC073924.1 - 11 1071
BT006920.1 - 5 522
CR456975.1 - 5 522
U86602.1 - 5 664

View Full Product Details