Gene Transcripts

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
Chromosome Location
Chr. 3: 108296490 - 108378285 on Build GRCh38
UniGene ID
Hs.225968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282556.1 NP_001269485.1 5 710
NM_001282557.1 NP_001269486.1 5 641
NM_001282558.1 NP_001269487.1 6 788
NM_001282559.1 NP_001269488.1 6 526
NM_007072.3 NP_009003.1 5 901
XM_005247080.3 XP_005247137.1 6 771
XM_011512362.1 XP_011510664.1 6 771
XM_011512363.1 XP_011510665.1 5 701
XM_011512364.1 XP_011510666.1 5 652
XM_011512367.2 XP_011510669.1 6 771
XM_017005642.1 XP_016861131.1 5 701
GenBank mRNA AF126162.1 - 5 641
AK000692.1 - 4 636
AK027132.1 - 2 194
AK296644.1 - 6 469
AK315109.1 - 6 733
BC035971.1 - 5 901 4606764

View Full Product Details