Gene Transcripts
- Gene Symbol
- NISCH
- Entrez Gene ID
- 11188
- Gene Name
- nischarin
- Gene Aliases
- I-1, IR1, IRAS, hIRAS
- Chromosome Location
- Chr. 3: 52455508 - 52493072 on Build GRCh38
- UniGene ID
- Hs.435290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276293.1 |
NP_001263222.1
|
9, 10 |
1132 |
|
NM_001276294.1 |
NP_001263223.1
|
9, 10 |
1132 |
|
NM_007184.3 |
NP_009115.2
|
9, 10 |
1132 |
|
XM_005264839.2 |
XP_005264896.1
|
8, 9 |
905 |
|
GenBank mRNA |
AB023192.1 |
- |
10, 11 |
1070 |
|
AF082516.1 |
- |
9, 10 |
1024 |
|
AK001505.1 |
- |
7 |
1245 |
|
AK027428.1 |
- |
9, 10 |
1010 |
|
AK291398.1 |
- |
9, 10 |
1036 |
|
AK302441.1 |
- |
1 |
20 |
|
AK315001.1 |
- |
9, 10 |
1036 |
|
BC038102.2 |
- |
9, 10 |
1036 |
5268247 |
BC054494.1 |
- |
9, 10 |
1030 |
6195532 |
BC056900.1 |
- |
9, 10 |
1002 |
6143316 |
View Full Product Details