Gene Transcripts

Gene Symbol
SHANK2
Entrez Gene ID
22941
Gene Name
SH3 and multiple ankyrin repeat domains 2
Gene Aliases
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
Chromosome Location
Chr. 11: 70467856 - 71252724 on Build GRCh38
UniGene ID
Hs.268726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012309.4 NP_036441.2 17 2245
NM_133266.4 NP_573573.2 3 442
NR_110766.1 3 442
XM_005277932.3 XP_005277989.1 8 1250
XM_017017387.1 XP_016872876.1 18 2350
XM_017017388.1 XP_016872877.1 18 2372
XM_017017389.1 XP_016872878.1 18 2545
XM_017017390.1 XP_016872879.1 5 3483
GenBank mRNA AB208025.1 - 17 2211
AB208026.1 - 8 1033
AB208027.1 - 3 420
AF141901.1 - 3 334
BC093885.1 - 3 442
BC112097.1 - 3 442
BC114484.1 - 3 364
BC143389.1 - 3 442

View Full Product Details